Z147311
Lecture-bottle adapter, NPT to NPT
brass
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.51
Polecane produkty
Materiały
brass
wlot i wylot gwint zewn. × gwint zewn.
1/4 in. × 1/4 in.
Opis ogólny
For connection of valves, regulators, and filters to cylinder valves.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)