Z135801
Scienceware® POXYGRID® test-tube rack
Holds 24 x 15-16 mm tubes
Synonim(y):
uchwyt rurki
About This Item
Kod UNSPSC:
41122800
NACRES:
NB.35
Polecane produkty
Materiały
epoxy (coated steel)
Właściwości
Holds 24 x 15-16 mm tubes
opakowanie
pack of 1 ea
producent / nazwa handlowa
Bel-Art F18788-1601
dł. × szer. × wys.
124 mm × 89 mm × 67 mm
Powiązane kategorie
Opis ogólny
Welded steel wire is coated with heat-cured, thermosetting epoxy to protect from rust and corrosion. The coating is unaffected by most organic solvents, weak acids and alkalis, and has excellent abrasion resistance and adhesion to steel substrate.
Temp. range: -370 to 250°F (-233 to 121°C). Has square holes, three decks, slotted-mesh bottom deck and reinforced edges.
Temp. range: -370 to 250°F (-233 to 121°C). Has square holes, three decks, slotted-mesh bottom deck and reinforced edges.
Informacje prawne
Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z135801-1EA | 4061832256863 |