Kluczowe dokumenty
CAD4181
Stainless steel 304 syringe needle
gauge 21, L 6 in.
About This Item
Kod UNSPSC:
42142600
NACRES:
NB.03
opis
Standard Hub with 304 Stainless Steel Tubing
opakowanie
pack of 12 ea
producent / nazwa handlowa
Cadence Science 4181
Długość
6 in.
grubość
21
Opis ogólny
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)