HPLCによる血清中の三環系抗うつ薬の分析、Discovery® C18、SPE処理、Discovery® DSC-18使用
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
SPE tube/cartridge
Discovery DSC-18, 100 mg/1 mL (52602-U)
column
Discovery C18, 15 cm × 4.6 mm, 5 μm preceded by a 2 cm C18 guard column and 0.5 μm frit filter (504955)
mobile phase
[A] acetonitrile: [B] methanol: [C] 25 mM potassium phosphate, monobasic (pH 7 with triethlylamine) (45:25:30, A:B:C)
flow rate
1 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
50 μL, diluted porcine serum extract