HPLCによるピンドロールエナンチオマーの分析、 CHIROBIOTIC® V
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine; (99.98:0.01:0.01, A:B:C)
flow rate
2 mL/min
column temp.
23 °C
detector
UV, 254 nm