HPLCによるセロトニン代謝物の分析、Ascentis® C18
材料
Standard
分析カラム
column
Ascentis C18, 15 cm x 4.6 mm I.D., 5 μm particles (581324-U)
mobile phase
[A] 10 mM formic acid/acetate (pH 3.0): [B] methanol; (80:20, A:B)
flow rate
0.7 mL/min
column temp.
35 °C
detector
ESI (+); overlay of extracted ion chromatograms of individual (M+H)+ species
injection
10 μL
sample
25 mg/L in 90:10, 10 mM formic acid/acetate (pH 3.0):methanol