HPLCによるワルファリン抗凝固剤の分析、Ascentis® C18
材料
分析カラム
column
Ascentis C18, 15 cm x 4.6 mm I.D., 5 μm particles (581324-U)
mobile phase
[A] 20 mM phosphoric acid (pH 2.0, unadjusted): [B] methanol; (30:70, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 280 nm
injection
10 μL
sample
50 μg/mL each in mobile phase