Merck
Z763462
MAGic Clamp universal platform for flasks and tube racks
L × W 18 in. × 18 in.
別名:
magnetic clamps, orbital shaker platforms
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41122413
NACRES:
NB.77
メーカー/製品名
Benchmark Scientific H1010-MR
L × W
18 in. × 18 in.
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The convenient MAGic Clamp platforms (large or small) are available for use with Erlenmeyer flasks and test tube racks. The unique magnetic attachment method is widely recognized as the easiest way to instantly change between flask clamps of different sizes.
法的情報
MAGic Clamp is a trademark of Benchmark Scientific, Inc.
文書ライブラリにアクセスする
この製品を見ている人はこちらもチェック
Slide 1 of 3
1 of 3
Incu-Shaker™ 10 L AC/DC input 115 V AC
Z765686
Incu-Shaker 10 L
Shaker platform orbital shaker platform, size 21.7 mm ( 7/8 in.) , rotating orbit diameter
Z363618
Shaker platform