Merck
Z659703
Bandelin SONOREX 超音波バスアクセサリー
tank lid for Z659673 and Z659029
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41103706
NACRES:
NB.74
物質
stainless steel
包装
pack of 1 ea
メーカー/製品名
Bandelin D 255
利用可能性
not available in USA and Canada
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Stainless steel lid for an ultrasonic bath protects the bath liquid from external contamination and prevents unpleasant odors from the cleaning or disinfection agents added. In addition, the noise emitted when the ultrasonic cleaner is in operation, which may be perceived as annoying, is reduced. The lids are designed in such a way that any condensation that occurs is directed back into the bath.
特徴および利点
  • protection the bath fluid from contamination
  • sound-absorbing
  • shortens the heating phase
法的情報
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
最新バージョンのいずれかを選択してください:
試験成績書(COA)