詳細
Compact, convenient 80 well racks designed to keep your tubes neatly and efficiently organized. Theses autoclavable polypropylene racks are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification. The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes.
Tube Rack, fraction collector type, 80 x 1.5mL/2mL, green, pack of 5
特徴および利点
- 80 well (5 rows of 16) for 1.5 to 2.0mL microtubes
- Rainbow pack available — 5 colors
- Two frosted marking areas, on the bottom and side
- Raised numbers beside each well
- Recessed well bottoms for improved tube stability
法的情報
MTC is a trademark of MTC Bio
最新バージョンのいずれかを選択してください:
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
MTCR1040R | 4065270485892 |
MTCR1040R-5EA | 4065271097117 |