Merck
EPM1289-0060
Test Tube Rack, for Eppendorf® New Brunswick Innova® and Eppendorf® Excella® Shakers
size 26‑30 mm medium, holds 16 tubes
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41122804
NACRES:
NB.73
サイズ
26‑30 mm medium
360x80x129 mm
特徴
holds 16 tubes
メーカー/製品名
Eppendorf® M1289-0060 (Global)
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
法的情報
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG
最新バージョンのいずれかを選択してください:
試験成績書(COA)