Merck
Z635545
Swiss Boy lab jacks
load ≤80 kg (with two plates, middle position), platform L × W 200 mm × 200 mm, Model 115
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41122800
NACRES:
NB.35
メーカー/製品名
Grauer
パラメーター
≤80 kg load (with two plates, middle position)
76-280 mm vertical limit
プラットフォームL × W
200 mm × 200 mm
gold
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
These high quality Swiss made jacks have massive cast aluminum frames with a chemically resistant anodised finish. Swiss boy jacks are designed to carry heavy loads safely. Available either in standard size or with extended carrying plates for large pots or pans. Units carrying more that 90kg are also available with a motorized lift adjustment for enhanced convenience. The max load is for the middle position. Supplied with top and bottom plates.
Products with motorized jacks are available in Europe only.
Jack can be raised with load in place.
最新バージョンのいずれかを選択してください:
試験成績書(COA)
文書ライブラリにアクセスする
この製品を見ている人はこちらもチェック
Slide 1 of 4
1 of 4
Swiss Boy lab jacks load ≤30 kg (middle position), platform L × W 80 mm × 75 mm, Model 105
Z635324
Swiss Boy lab jacks
Support jack platform L × W 10 cm × 10 cm
Z565814
Support jack
Support jack platform L × W 20 cm × 20 cm
Z565830
Support jack
Global Trade Item Number
カタログ番号GTIN
Z635545-1EA4061833581582