ジョイント
joint: ST/NS29/32
外径
60 mm
表面積
1230 cm2
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Excellent for lower boiling solvents.
Features:
• Triple-jacketing provides maximum cooling surface area
• Does not flood as readily during reflux surges
• Minimizes loss of volatile components
• Highly efficient while extremely compact
Features:
• Triple-jacketing provides maximum cooling surface area
• Does not flood as readily during reflux surges
• Minimizes loss of volatile components
• Highly efficient while extremely compact
法的情報
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
最新バージョンのいずれかを選択してください:
Global Trade Item Number
カタログ番号 | GTIN |
---|---|
Z517445-1EA | 4061833640715 |