Merck
DWK14395-250
KIMBLE® KIMAX® Media bottle
with cap, round transparent borosilicate glass bottle, bottle capacity (250 mL), non-sterile
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121800
NACRES:
NB.43
物質
blue polypropylene cap
round transparent borosilicate glass bottle
無菌性
non-sterile
特徴
with cap
包装
case of 10 ea
メーカー/製品名
Kimble® 14395-250
ボトルの最大容量
(250 mL)
フィッティング
GL 45 neck
GL 45
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
GL 45 Media Bottles ideal for general laboratory use including mixing, storing or transporting culture media, chemicals or solvents.
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
特徴および利点
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
法的情報
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)