Documenti fondamentali
SYND261024
Synthware short path distillation head
joint: ST/NS 24/40, thermometer joint: ST/NS 10/18, immersion depth 25 mm
Sinonimo/i:
Synthware D261024, distilling head
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
Giunto
thermometer joint: ST/NS10/18
joint: ST/NS24/40
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
D261024
Prof. immersione
25 mm
Categorie correlate
Descrizione generale
Synthware distillation head features a close coupled condenser and vacuum connection to offer a short condensate travel path. One-piece design for easy setup. For the best results, use the D272440 distillation receiver. It is designed to be used with a 25 mm immersion thermometer.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)