Documenti fondamentali
SYND214300
Synthware vacuum jacketd Vigreux distillation head
joint: ST/NS 24/40, thermometer joint: ST/NS 10/18, column H 300 mm, immersion depth 25 mm
Sinonimo/i:
Synthware D214300, short path head, Vigreux distilling head
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.51
Giunto
thermometer joint: ST/NS10/18
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware D214300
Alt. colonna
300 mm
Prof. immersione
25 mm
Descrizione generale
Vacuum jacketed Vigreux distillation head having a sealed on condenser, braced for strength. Vacuum adapter has an inner joint and a sealed in drip tip. To be used with a 25 mm immersion thermometer
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)