Documenti fondamentali
SYND121014
Synthware vacuum jacketed short path distillation head
joint: ST/NS 14/20, thermometer joint: ST/NS 10/18, immersion depth 25 mm
Sinonimo/i:
Synthware D121014, Distilling head
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
thermometer joint: ST/NS10/18
joint: ST/NS14/20
Produttore/marchio commerciale
D121014
Prof. immersione
25 mm
Descrizione generale
Synthware vacuum-jacketed design for enhanced efficiency. When a thermometer is inserted at the top of this apparatus, the bulb and stem act as column packing. The close-coupled condenser and vacuum connection provide an extremely short condensate travel path. This apparatus is compatible with a 25 mm immersion thermometer and a D27 distillation receiver.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)