主要文件
XX42PFK01
Millipore
Pellicon® Mini Holder Fitting Kit Polypropylene
登录查看公司和协议定价
About This Item
分類程式碼代碼:
23151800
eCl@ss:
32031690
材料
polypropylene
品質等級
製造商/商標名
Pellicon®
一般說明
The Pellicon® Mini Holder Fitting Kit Polypropylene is an accessory to the Pellicon® Mini Cassette Holder which has a simple design which makes process development and small scale pharmaceutical manufacturing simple and efficient.
The Pellicon® Mini holder operates one to three Pellicon® 2 Mini cassettes in parallel or Pellicon® 3 cassettes. This sanitary holder is tightened with a small torque wrench to compress the cassettes between a manifold plate that conveys fluids in and out of the cassettes and an end plate that seals the cassettes together.
The Mini holder is designed for process development and small volume pharmaceutical manufacturing.
特點和優勢
The Mini holder and Pellicon® Mini Holder Fitting Kit Polypropylene is designed for process development and small volume pharmaceutical manufacturing.
法律資訊
Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany