Documenti fondamentali
Z227331
Frit for Supelco® Filter
stainless steel, pore size 2.0 μm, pkg of 5 ea
About This Item
Codice UNSPSC:
41115700
NACRES:
SB.52
Materiali
stainless steel
Confezionamento
pkg of 5 ea
Dimensione pori
2.0 μm
Applicazioni
This product may be used to protect HPLC columns containing 5 μm or larger particles against particulate matter that gets accumulated during the chromatographic operations.
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Ricambio
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 3
1 of 3
Column Frits diam. 1/8 in., pore size 0.5 μm
59037
Column Frits
PEEK Inline Microfilter
502707
PEEK Inline Microfilter
Articoli
Identify and correct common HPLC problems to restore column performance. Get help with peak problems (peak splitting, peak tailing, peak fronting, etc.), retention time, purge noise, and loss of resolution with example chromatograms.
Global Trade Item Number
SKUGTIN
Z2273314061833461525