Key Documents
Z227323
Supelco® Precolumn Filter
Direct-connect, PEEK
Sinonimo/i:
TM=["Supelco"] Guard Column Filter
About This Item
Codice UNSPSC:
41115700
NACRES:
SB.52
Materiali
PEEK
Descrizione
starter kit
Parametri
100 °C max. temp.
5000 psi max. pressure
Foro
0.015 in.
Descrizione generale
Direct-connect; protects analytical and guard columns. Our precolumn filter can be connected directly, hand-tightened, into any HPLC column or guard column that has Valco-compatible end fittings. PEEK cap and body, 2 μm stainless steel frit. For a metal-free system, order PEEK (biocompatible) replacement frits (57430-U).
Prodotti consigliati
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Note legali
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Raccomandato
N° Catalogo
Descrizione
Determinazione del prezzo
Ricambio
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 1
1 of 1
EXP® Pre-Column Filter holder with EXP titanium hybrid ferrule, pack of 1 sets
51163-U
EXP® Pre-Column Filter
Articoli
Identify and correct common HPLC problems to restore column performance. Get help with peak problems (peak splitting, peak tailing, peak fronting, etc.), retention time, purge noise, and loss of resolution with example chromatograms.