Documenti fondamentali
Z765554
Tube rack for Benchmixer XL multi-tube vortexer
Holds 50 x 1.5/2.0 mL tubes
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.77
Caratteristiche
Holds 50 x 1.5/2.0 mL tubes
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Benchmark Scientific BV1010-1520
Descrizione generale
Benchmixer XL vortexer head adapters to hold multiple tubes.
This rack enables the use of 50 x 1.5/2.0 mL tubes in the BenchMixer XL heavy-duty hands-free mixing vortex mixer. Ideal for the QuEChERS pesticide analysis method.
Note legali
Benchmixer is a trademark of Benchmark Scientific, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Articoli
Extraction of Permethrin Pesticides from Spinach Using QuEChERS Methodology with Automated Shaking