Z258423
Vortex-Genie® 2 mixer
AC/DC input 220 V AC, Schuko plug
Sinonimo/i:
Laboratory vortex mixer, Vortex mixer
About This Item
Codice UNSPSC:
41103800
NACRES:
NB.73
Prodotti consigliati
Caratteristiche
CE compliant
CSA approved
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Scientific Industries SI-0256
Scientific Industries
Categorie correlate
Descrizione generale
Mixes liquids quickly and thoroughly with vortex action. Variable speed control allows precise mixing range. The unit contains a standard 3 in. platform head for flasks and beakers, and a cup head for single tube mixing. Accessories for special applications are sold separately. Speed: 600 to 3,200 rpm. Max. load: 1 lb.
- Variable speed control
- Shaking or vortex action
- Continuous or touch-control mixing
Note legali
Vortex-Genie is a registered trademark of Scientific Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)