Documenti fondamentali
Z258458
Sigma-Aldrich
Head set for Vortex-Genie® 2
Large sample
Sinonimo/i:
Head Set for Vortex, Vortex Adapter, Vortex-Genie Head Set
About This Item
Codice UNSPSC:
41103814
NACRES:
NB.73
Produttore/marchio commerciale
Scientific Industries
Descrizione generale
For continuous vortexing of beakers, flasks, and tubes. Includes a recessed platform, elastic flask harness, and an assortment of tube inserts (accepts 10 to 37mm diam. tubes).
Note legali
Vortex-Genie is a registered trademark of Scientific Industries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Global Trade Item Number
SKUGTIN
Z258458-1PAK4061833568958