Key Documents
Z376825
Sigma-Aldrich
Scintillation vials, general purpose
polypropylene
Sinonimo/i:
Scintillation Vials
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.11
Materiali
polypropylene
Sterilità
non-sterile
Confezionamento
pkg of 1000 ea
Produttore/marchio commerciale
Simport S207-5
Misura flaconcino
16 mm × 57 mm
Volume
6.5 mL
Descrizione generale
General purpose vials can be used for liquid scintillation counting, gamma counting, chromatography, sample storage, and culturing. Fits LKB and Packard Varisette counters. Vials are securely sealed by snapping the cap on. Cap removal requires a simple 1/4 twist, reducing the chance of spillage associated with pull of caps. Polyethylene vials are supplied with polypropylene caps. Polypropylene vials are supplied with polyethylene caps.