Documenti fondamentali
TMO362150-5200
Nalgene® red caps
polypropylene cap, diam. × thread 22 mm × 20-415 mm, case of 2000 ea
Sinonimo/i:
bottle cap, bottle closure
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
polypropylene cap
red cap
Confezionamento
case of 2000 ea
Produttore/marchio commerciale
Nalgene 362150-5200
Diam. × filettatura
22 mm × 20-415 mm
Descrizione generale
Eliminate spills with Thermo Scientific Nalgene Colored Polypropylene Closures. These one-piece linerless closures fit Nalgene narrow-mouth 4mL bottles. They feature an integrally molded seal ring and a well-defined closure knurl that makes gripping and capping easy.
Note legali
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)