Documenti fondamentali
Z106402
Supelco
Black plastic caps
Solid top, white rubber liner, thread for 13-425, pkg of 200 ea
Sinonimo/i:
Black Polymer Caps
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.11
Materiali
black phenolic cap
white rubber liner
Caratteristiche
closure type black phenolic solid top cap
Confezionamento
pkg of 200 ea
Raccordi
thread for 13-425
Descrizione generale
It is autoclavable, easy sterilization, is used as a closure type for the containers used to contain substances in order to prevent contamination.
These autoclavable caps are designed to ensure the ultimate in product safety.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z106402-200EA4061837589300