Z188832
Black plastic caps
Solid top, PTFE-faced rubber liner, thread for 18-400, pkg of 200 ea
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.11
Prodotti consigliati
Materiali
PTFE-faced rubber liner
black phenolic cap
Caratteristiche
closure type black phenolic solid top cap
Confezionamento
pkg of 200 ea
Raccordi
thread for 18-400
Descrizione generale
These autoclavable caps are designed to ensure the ultimate in product safety.
Applicazioni
- Marine litter accumulation along the Bulgarian Black Sea coast: Categories and predominance.: This research categorizes and quantifies the types of marine litter found along the Bulgarian Black Sea coast, with a particular focus on the prevalence of black plastic caps. The findings underscore the necessity for targeted clean-up efforts and policy interventions to address specific types of marine debris (Simeonova & Chuturkova, 2019).
Prodotti correlati
N° Catalogo
Descrizione
Determinazione del prezzo
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z188832-1PAK | 4061837617256 |