Key Documents
EP5810733006
Eppendorf® Replacement cap sealing for aerosol-tight caps
for rotor A-4-81, S-4x500, pkg of 4 ea
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.82
Caratteristiche
for rotor A-4-81, S-4x500
Confezionamento
pkg of 4 ea
Produttore/marchio commerciale
Eppendorf® 5810733006
Note legali
Eppendorf is a registered trademark of Eppendorf AG