Documenti fondamentali
CLS430597
Corning® non-treated culture dishes
D × H 150 mm × 25 mm
Sinonimo/i:
cell culture dishes, cell culture plates, culture dish
About This Item
Codice UNSPSC:
41121812
NACRES:
NB.13
Materiali
polystyrene
round
Sterilità
sterile
Confezionamento
sleeve of 5
Produttore/marchio commerciale
Corning 430597
D × alt.
150 mm × 25 mm
Area superficiale
148 cm2
Descrizione generale
  • Manufactured from optically-clear virgin polystyrene
  • Not treated for cell attachment, for applications where cell attachment is not desired
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange
The 150 and 245 mm culture dishes make excellent carriers and incubator trays for 35 and 60 mm dishes. This avoids spills and lowers the risk of contamination.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 2
1 of 2
Global Trade Item Number
SKUGTIN
CLS430597-60EA4061833887288