CLS431110
Corning® square bioassay dishes
without handles, TC treated, square
Sinonimo/i:
cell culture dishes, cell culture plates, culture dish
About This Item
Prodotti consigliati
Materiali
polystyrene
square
Sterilità
sterile; γ-irradiated
Confezionamento
sleeve of 4 x 4
Produttore/marchio commerciale
Corning 431110
Altezza
25 mm
Lungh. × largh.
245 mm × 245 mm
Area superficiale
500 cm2
Descrizione generale
- Corning® square bioassay dishes are made from optically clear polystyrene.
- Stacking bead design facilitate secure stacking and prevents slipping.
- Compatible with automated colony picking instruments.
- Certified nonpyrogenic dishes are supplied with lids.
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)