Documenti fondamentali
CLS430180
Corning® polypropylene beaker
sterile, size 250 mL , beaker, container only, case of 500
Sinonimo/i:
plastic beakers, polypropylene beakers, storage containers
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.13
Materiali
natural polypropylene (beaker)
orange high-density polyethylene (snap cap)
Sterilità
sterile
sterile
Caratteristiche
container only
Confezionamento
pack of 20
case of 500
Produttore/marchio commerciale
Corning 430180
Misura
250 mL , beaker
Descrizione generale
Corning flexible polypropylene container with snap-on polyethylene lid serves as a beaker or storage container.
  • Container is graduated in both millilitres and ounces
  • Certified nonpyrogenic
Altre note
Browse all plastic beakers
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
CLS430180-500EA4061831708448