CLS430181
Corning® polypropylene beaker
lid only, for 250 mL beaker, sterile, 500/cs
Sinonimo/i:
Corning container
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.13
Prodotti consigliati
Materiali
natural polypropylene (beaker)
orange high-density polyethylene (snap cap)
Sterilità
sterile
Confezionamento
pack of 20
case of 500
Produttore/marchio commerciale
Corning 430181
Categorie correlate
Descrizione generale
Corning flexible polypropylene container with snap-on polyethylene lid serves as a beaker or storage container.
- Container is graduated in both millilitres and ounces
- Certified nonpyrogenic
These replacement snap-on polyethylene lids fit on the Corning 250mL containers (CLS430179 and CLS430180) and are certified nonpyrogenic.
Note legali
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
CLS430181-500EA | 4061831708455 |