CLS3200
Corning® CellCube® Module 10-Layer with 8500 cm2 growth surface
polystyrene (growth surface), sterile, case of 2
Sinonimo/i:
cell culture vessel
About This Item
Codice UNSPSC:
41122104
NACRES:
NB.13
Prodotti consigliati
Materiali
polystyrene (growth surface)
Sterilità
sterile
Caratteristiche
graduations: no
Confezionamento
pack of 1
case of 2
Produttore/marchio commerciale
Corning 3200
Area superficiale
8500 cm2
Volume d’esercizio
600 mL
Descrizione generale
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth.
These CellCube Modules provide:
Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth.
These CellCube Modules provide:
- a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
- 8,500cm2 polystyrene growth surface optimally treated for the growth of attachment dependent cells.
- a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen.
- an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment
Note legali
CELLCUBE is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CLS3200-2EA | 4061833883723 |