Key Documents
AXYRESSW12HPSI
AxyVoir reservoir
12 bottom, high profile single-well, individually wrapped, sterile
Sinonimo/i:
storage reservoir
About This Item
Codice UNSPSC:
41121522
NACRES:
NB.22
Sterilità
sterile
Produttore/marchio commerciale
Axygen Inc
Corning