AXYRESMW12HP
Reservoir
12 channel, high profile multiwells, size 7 mL, 25 ea of
Sinonimo/i:
storage reservoir
About This Item
Codice UNSPSC:
41121522
NACRES:
NB.22
Prodotti consigliati
Produttore/marchio commerciale
Axygen Inc RES-MW12-HP
Corning
Confezionamento
25 ea of
Misura
7 mL
Categorie correlate
Descrizione generale
- Axygen® reagent reservoir is a multiple-well, high-profile reservoir manufactured using chemically resistant polypropylene.
- Ideal for both manual and automated applications.
- Designed with standard ANSI/SLAS footprint dimensions for microplates.
- Features 12-Channel trough - extended length of the bottom surface.
- Designed for 12-channel applications.
Note legali
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)