BAF378820000
Three-in-one multi-channel pipette reservoir
pack of 10 ea
About This Item
Codice UNSPSC:
41121522
NACRES:
NB.35
Prodotti consigliati
Materiali
V-bottom bottom
polypropylene
white
Sterilità
non-sterile
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
Bel-Art F37882-0000
Capacità
60 mL
Categorie correlate
Descrizione generale
Suitable for dispensing into multi-tube blocks or microwell plates with a multichannel pipette, this handy reservoir is actually 3 reservoirs in a single unit:
1. A graduated single solution reservoir with graduations at 10, 25, 40 and 50 mL.
2. A reservoir with 8 separate 5 mL (max. capacity) solution channels.
3. A reservoir with 12 separate 5 mL (max. capacity) solution channels.
All channels of the multi-channel reservoirs are numbered for easy reagent identification. Compatible with all makes of multi-channel pipettors. Sterilizable.
1. A graduated single solution reservoir with graduations at 10, 25, 40 and 50 mL.
2. A reservoir with 8 separate 5 mL (max. capacity) solution channels.
3. A reservoir with 12 separate 5 mL (max. capacity) solution channels.
All channels of the multi-channel reservoirs are numbered for easy reagent identification. Compatible with all makes of multi-channel pipettors. Sterilizable.
- Three built in reservoirs in one unit: one single chamber reservoir, one 8-channel reservoir and a 12 channel reservoir
- Polypropylene construction
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)