PUMP05601
Recirculation Pump
AC/DC input 120 V
About This Item
Codice UNSPSC:
41104200
NACRES:
SB.51
Prodotti consigliati
Ingresso CA/CC
120 V, 60 Hz
Confezionamento
pkg of 1 unit
Compatibilità
for use with tanks
Descrizione generale
Recirculation Pump is a water purification system accessory. The pump recirculates stored water to maintain pure water quality.
For use with 30, 60, and 100 L polyethylene (PE) reservoirs to protect stored water quality.
For use with 30, 60, and 100 L polyethylene (PE) reservoirs to protect stored water quality.