Z677507
Camapren™ chloroprene flock lined gloves
size L
Sinonimo/i:
reusable glove
About This Item
Prodotti consigliati
Materiali
red (chloroprene, natural latex)
red gloves
Misura
L
Sterilità
non-sterile
Caratteristiche
CE compliant
disposable
latex free: no
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
standard type EN 421
Confezionamento
package of 100 ea
Produttore/marchio commerciale
KCL 722 - 09
KCL 722
Lungh. × spessore
300 mm × 0.6 mm
Descrizione generale
The cotton flock lining improves comfort. Altogether, this is a product range for excellent comfort and wearing characteristics, reliable, economical and popular with users.
- Ergonomic fit and high elasticity ensures good sensitivity
- Grooved palm for excellent grip when wet
- High mechanical resistance
- Cotton flock lining improves comfort
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- Protection from radioactive contamination
- CE compliant
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z677507-100EA | 4061833069707 |