Documenti fondamentali
Z677566
Lapren latex/chloroprene flock lined gloves
size L
Sinonimo/i:
chemically resistant glove
About This Item
Codice UNSPSC:
46181504
NACRES:
NB.35
Materiali
green (chloroprene, natural latex)
green gloves
Misura
L
Sterilità
non-sterile
Caratteristiche
CE compliant
disposable
latex free: no
powder-free
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
Confezionamento
package of 100 ea
Produttore/marchio commerciale
KCL 706 - 09
KCL 706
Lungh. × spessore
310 mm × 0.6 mm
Descrizione generale
Lapren incorporates a high quantity of polychloroprene into latex to improve chemical resistance. A mixture of raw materials ensures a high elasticity as well as good tensile strength and tear resistance. The palm grip surface ensures a firm grip even in wet working environments without loss of the high degree of sensitivity.
With cotton flock lining, stretched cuff and anatomical design, this product meets all requirements of comfort and hygiene.
  • Ergonomic fit and high elasticity ensures good sensitivity
  • Profiled palm for firm grip when wet
  • High mechanical resistance
  • Cotton flock lining improves comfort
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • CE compliant
Note legali
Lapren is a trademark of KCL GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z677566-100EA4061833069806