Z648280
Spinbar® magnetic stirring bar garage
keep all your magnetic stirring bars and accessories organized and accessible
About This Item
Codice UNSPSC:
41103806
NACRES:
NB.72
Prodotti consigliati
Materiali
epoxy-coated
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art F37790-0000
Misura
25.6 cm (10.3 in.) × 10 cm (4 in.) × 31 cm (12.3 in.)
Categorie correlate
Descrizione generale
Keep all your magnetic stirring bars and accessories organized and accessible with the Spinbar magnetic stirring bar garage.
Multiple sizes and shapes of magnetic stirring bars from 159 mm (6") to tiny 10 mm (0.5") bars can be stored on the dedicated rack and free stands on the bench or can be mounted to a vertical surface (mounting hardware not included).
Tight wire spacing in the upper section of the rack holds smaller magnetic stirs bars, keeping them separate from larger bars stored below. A lower wire shelf holds small lab tools and prevents stirring bars from accidental falls.
Multiple sizes and shapes of magnetic stirring bars from 159 mm (6") to tiny 10 mm (0.5") bars can be stored on the dedicated rack and free stands on the bench or can be mounted to a vertical surface (mounting hardware not included).
Tight wire spacing in the upper section of the rack holds smaller magnetic stirs bars, keeping them separate from larger bars stored below. A lower wire shelf holds small lab tools and prevents stirring bars from accidental falls.
Note legali
Spinbar is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z648280-1EA | 4061832266084 |