Z646199
Spinbar® restrainer
retrieve magnetic stirring bars from vessels, easily, pkg of 1 ea
About This Item
Codice UNSPSC:
41103806
NACRES:
NB.72
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Bel-Art F37786-1000
Categorie correlate
Descrizione generale
Easily retrieve magnetic stirring bars from vessels with the handheld Spinbar Restrainer. This restrainer stops losses of magnetic stirring bars headed down lab drains and eliminates the splashing hazard when pouring out of mixing containers. The tool fits comfortably in one hand and when placed next to the outside of a mixing container, readily attracts and holds stirring bars in place.
As containers are emptied, the stirring bar is restrained and can be safely removed from the vessel. The Spinbar Restrainer body is molded of easy to clean high impact polystyrene. It contains a powerful magnet that attracts and holds magnetic stirring bars up to 80 mm long.
As containers are emptied, the stirring bar is restrained and can be safely removed from the vessel. The Spinbar Restrainer body is molded of easy to clean high impact polystyrene. It contains a powerful magnet that attracts and holds magnetic stirring bars up to 80 mm long.
Note legali
Spinbar is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z646199-1EA | 4061832265988 |