Z422274
Aldrich® triple-jacket coil condenser
joint: ST/NS 24/40
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Giunto
joint: ST/NS24/40
D.E.
60 mm
Area superficiale
1230 cm2
Descrizione generale
Excellent for lower boiling solvents.
Features:
• Triple-jacketing provides maximum cooling surface area
• Does not flood as readily during reflux surges
• Minimizes loss of volatile components
• Highly efficient while extremely compact
Features:
• Triple-jacketing provides maximum cooling surface area
• Does not flood as readily during reflux surges
• Minimizes loss of volatile components
• Highly efficient while extremely compact
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Global Trade Item Number
SKU | GTIN |
---|---|
Z422274-1EA | 4061833640593 |