Documenti fondamentali
SYNC141120
Synthware Graham condenser
joint: ST/NS 14/20, hose size 8 mm
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C141120
Dimens. tubo
8 mm
Categorie correlate
Descrizione generale
Synthware Graham condenser features a 120 mm jacketed length for superior cooling efficiency. With standardized 14/20 upper and lower joints, it easily fits various laboratory glassware. Made from durable borosilicate glass.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)