Documenti fondamentali
SYNC144300
Synthware Graham condenser
joint: ST/NS 24/40, hose size 10 mm
Sinonimo/i:
Synthware C179200, distillation condenser, Inland Revenue condenser
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C144300
Dimens. tubo
10 mm
Categorie correlate
Descrizione generale
Synthware Graham condenser features a 300 mm jacketed length for superior cooling efficiency. With standardized 24/40 upper and lower joints, it easily fits various laboratory glassware. Made from durable borosilicate glass.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)