Z256811
Ace double-jacketed coiled condenser with Ace-Thred™ hose connections
jacket L 250 mm
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Giunto
joint ST/NS24/40
Produttore/marchio commerciale
Ace Glass 5956143
Lunghezza camicia
250 mm
Categorie correlate
Descrizione generale
With #7 Ace-Thred hose connections for 1/4 in. I.D. tubing. Extra cooling capacity for volatile materials. Features threaded inlet/outlet connections that incorporate UHDPE (ultra-high-density PE) connectors and nylon bushings to make easy connect/disconnect of coolant tubing . The bushing compresses the connector and a silicone O-ring for a leaktight seal. Compact design retards diffusion.
Note legali
Ace-Thred is a trademark of Ace Glass, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z256811-1EA | 4061832363004 |