Documenti fondamentali
SYNC201080
Synthware jacketed reflux condenser
joint: ST/NS 14/10, jacket L 80 mm
Sinonimo/i:
Synthware C201080
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Giunto
joint: ST/NS14/10
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
C201080
Lunghezza camicia
80 mm
Categorie correlate
Descrizione generale
Synthware reflux condenser features a 80 mm jacketed length for superior cooling efficiency. I.D. of inner tube is designed large enough to accept a thermometer. Supplied with one holed compression cap and one Viton® O-ring. Made from durable borosilicate glass.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)