SYNC261100
Synthware™ coil-style reflux condenser
joint: ST/NS 14/20, hose size 8 mm
Sinonimo/i:
Synthware C261100, coil-type condenser
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Prodotti consigliati
Giunto
joint: ST/NS14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C261100
Dimens. tubo
8 mm
Categorie correlate
Descrizione generale
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 8mm or 10mm at the middle serration.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)