SYNC564125
Synthware™ coil-style reflux condenser with removable hose connections
joint: ST/NS 24/40, hose size 10 mm
Sinonimo/i:
Synthware C564125, coil-type condenser
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Prodotti consigliati
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware C564125
Dimens. tubo
10 mm
Categorie correlate
Descrizione generale
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)