Z116408
Double-ended needle
gauge 12, L 18 in.
About This Item
Codice UNSPSC:
41122002
NACRES:
NB.43
Prodotti consigliati
Confezionamento
pack of 6 ea
Produttore/marchio commerciale
Cadence Science
Lunghezza
18 in.
Gauge
12
Categorie correlate
Descrizione generale
304 SS. One noncoring tip, one flat-cut end.
Applicazioni
Double-ended needle has been used in the transfer of solution betweentwo flasks. It has also been used in a sequence alkylation reaction.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Ramón Guzmán-Mejía et al.
Nature protocols, 2(11), 2759-2766 (2007-11-17)
The use of microwave (MW) irradiation for the condensation reaction between acetophenone and alpha-phenylethylamine to prepare (R,R)-bis[alpha-phenylethyl]amine results in significantly reduced reaction times relative to the use of conventional heating. In this protocol, a secondary amine, (R,R)-bis(alpha-phenylethyl)amine is treated with
Global Trade Item Number
SKU | GTIN |
---|---|
Z116408-6EA | 4061832529929 |