TEM-FCF300NI
Formvar/Carbon Supported Nickel Grids
size 300 mesh, box of 25 ×
Sinonimo/i:
Formvar/Carbon 300 Mesh, Nickel, 25pk
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Prodotti consigliati
Confezionamento
box of 25 ×
Misura
300 mesh
Categorie correlate
Descrizione generale
A formvar coated TEM grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Thickness: 10nm Formvar and 1nm Carbon
Thickness: 10nm Formvar and 1nm Carbon
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
TEM-FCF300NI | 4061841758259 |